Assay Details
Gene Symbol: WRN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008252230.2 | XP_008250452.1 | 3 - 4 | 386 | 62 | |
XM_017339061.1 | XP_017194550.1 | 3 - 4 | 391 | 62 | ||
XM_017339066.1 | XP_017194555.1 | 3 - 4 | 390 | 62 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
