Assay Details
Gene Symbol: SS18L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_017343679.1 | XP_017199168.1 | 3 - 4 | 270 | 107 | |
XM_017343680.1 | XP_017199169.1 | 2 - 3 | 224 | 107 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
