Assay Details
Gene Symbol: IRAG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008259507.2 | XP_008257729.1 | 7 - 8 | 709 | 58 | |
XM_008259509.2 | XP_008257731.1 | 7 - 8 | 706 | 58 | ||
XM_008259510.2 | XP_008257732.1 | 6 - 7 | 514 | 58 | ||
XM_008259512.2 | XP_008257734.1 | 5 - 6 | 655 | 58 | ||
XM_017343328.1 | XP_017198817.1 | 6 - 7 | 740 | 58 | ||
XM_017343329.1 | XP_017198818.1 | 7 - 8 | 709 | 58 | ||
XM_017343330.1 | XP_017198819.1 | 2 - 3 | 141 | 58 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008259512.2, XM_017343330.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
