Assay Details
Gene Symbol: AREG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008267744.2 | XP_008265966.1 | 1 - 2 | 287 | 76 | |
XM_017347491.1 | XP_017202980.1 | 2 - 3 | 203 | 76 | ||
XM_017347492.1 | XP_017202981.1 | 2 - 3 | 183 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
