Assay Details
Gene Symbol: IL3RA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008248460.2 | XP_008246682.1 | 4 - 5 | 477 | 68 | |
XM_008248462.2 | XP_008246684.1 | 3 - 4 | 356 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() |
