Assay Details
Gene Symbol: CD70 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002723968.3 | XP_002724014.2 | 191 | 106 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_002723968.3 transcripts.
Set Membership: |
![]() ![]() ![]() |
