Assay Details
Gene Symbol: NEK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002717530.3 | XP_002717576.1 | 1 - 2 | 232 | 71 | |
XM_017347803.1 | XP_017203292.1 | 1 - 2 | 232 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
