Assay Details
Gene Symbol: PLA2G1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002719793.3 | XP_002719839.1 | 3 - 4 | 598 | 90 | |
GenBank mRNA | L11899.1 | - | 3 - 4 | 344 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
