Assay Details
Gene Symbol: CXCR3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002720089.3 | XP_002720135.1 | 1 - 2 | 236 | 94 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_002720089.3 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
