Assay Details
Gene Symbol: SOX10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002723532.3 | XP_002723578.1 | 217 | 87 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_002723532.3 transcripts.
Set Membership: |
![]() ![]() |
