Assay Details
Gene Symbol: RELL2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002710139.3 | XP_002710185.1 | 4 - 5 | 1211 | 61 | |
XM_008255149.2 | XP_008253371.1 | 4 - 5 | 1211 | 61 | ||
XM_017341141.1 | XP_017196630.1 | 4 - 5 | 1211 | 61 | ||
XM_017341143.1 | XP_017196632.1 | 4 - 5 | 1218 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
