Assay Details
Gene Symbol: SOD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082627.2 | NP_001076096.2 | 2 - 3 | 296 | 96 | |
XM_017347219.1 | XP_017202708.1 | 2 - 3 | 228 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() |
