Assay Details
Gene Symbol: GADD45A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261305.1 | NP_001248234.1 | 2 - 3 | 372 | 77 | |
GenBank mRNA | JU474506.1 | - | 2 - 3 | 372 | 77 | |
JV643929.1 | - | 2 - 3 | 425 | 77 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() |
