Assay Details
Gene Symbol: SNUPN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015142757.2 | XP_014998243.1 | 840 | 81 | ||
XM_028851160.1 | XP_028706993.1 | 608 | 81 | |||
XM_028851161.1 | XP_028706994.1 | 507 | 81 | |||
XM_028851162.1 | XP_028706995.1 | 636 | 81 | |||
GenBank mRNA | JU332393.1 | - | 4 - 5 | 476 | 81 | |
JU473849.1 | - | 4 - 5 | 448 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
