Assay Details
Gene Symbol: NSUN4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001108301.4 | XP_001108301.2 | 1274 | 87 | ||
XM_015138753.2 | XP_014994239.2 | 956 | 87 | |||
XM_015138762.2 | XP_014994248.2 | 958 | 87 | |||
GenBank mRNA | JU334441.1 | - | 4 - 5 | 782 | 87 | |
JU475536.1 | - | 4 - 5 | 838 | 87 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
