Assay Details
Gene Symbol: TEDC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001087250.4 | XP_001087250.1 | 546 | 87 | ||
XM_015144627.2 | XP_015000113.2 | 546 | 87 | |||
XM_028851958.1 | XP_028707791.1 | 546 | 87 | |||
XM_028851959.1 | XP_028707792.1 | 306 | 87 | |||
XM_028851960.1 | XP_028707793.1 | 300 | 87 | |||
GenBank mRNA | JU342081.1 | - | 480 | 87 | ||
JV613793.1 | - | 536 | 87 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() ![]() |
