Assay Details
Gene Symbol: SEC22A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194575.3 | NP_001181504.2 | 5 - 6 | 790 | 67 | |
XM_001112957.4 | XP_001112957.2 | 714 | 67 | |||
XM_015129917.2 | XP_014985403.1 | 800 | 67 | |||
XM_015129918.2 | XP_014985404.1 | 682 | 67 | |||
XM_028843388.1 | XP_028699221.1 | 656 | 67 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() ![]() |
