Assay Details
Gene Symbol: ZNF395 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015145045.2 | XP_015000531.2 | 3096 | 71 | ||
XM_015145046.2 | XP_015000532.2 | 3814 | 71 | |||
XM_015145048.2 | XP_015000534.2 | 364 | 71 | |||
XM_015145049.2 | XP_015000535.2 | 424 | 71 | |||
XM_015145050.2 | XP_015000536.2 | 436 | 71 | |||
XM_015145051.2 | XP_015000537.2 | 329 | 71 | |||
XM_015145052.2 | XP_015000538.2 | 375 | 71 | |||
XM_028852498.1 | XP_028708331.1 | 486 | 71 | |||
XM_028852499.1 | XP_028708332.1 | 410 | 71 | |||
XM_028852500.1 | XP_028708333.1 | 433 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() |
