Assay Details
Gene Symbol: RAP1A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261638.1 | NP_001248567.1 | 5 - 6 | 456 | 82 | |
GenBank mRNA | DQ148142.1 | - | 1 - 2 | 63 | 82 | |
JU335752.1 | - | 5 - 6 | 438 | 82 | ||
JU471889.1 | - | 5 - 6 | 442 | 82 | ||
JV045701.1 | - | 5 - 6 | 456 | 82 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() ![]() |
