Assay Details
Gene Symbol: C16H17orf75 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001257907.1 | NP_001244836.1 | 2 - 3 | 254 | 83 | |
GenBank mRNA | JU322894.1 | - | 2 - 3 | 254 | 83 | |
JU473688.1 | - | 2 - 3 | 245 | 83 | ||
JV044191.1 | - | 2 - 3 | 253 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() |
