Assay Details
Gene Symbol: MBD5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266707.1 | NP_001253636.1 | 7 - 8 | 1361 | 85 | |
XM_015110044.2 | XP_014965530.1 | 1239 | 85 | |||
XM_015110045.2 | XP_014965531.1 | 1109 | 85 | |||
XM_015110046.2 | XP_014965532.1 | 1238 | 85 | |||
XM_015110047.2 | XP_014965533.1 | 1236 | 85 | |||
XM_015110048.2 | XP_014965534.1 | 2094 | 85 | |||
XM_028830577.1 | XP_028686410.1 | 1386 | 85 | |||
XM_028830578.1 | XP_028686411.1 | 1565 | 85 | |||
XM_028830579.1 | XP_028686412.1 | 1677 | 85 | |||
XM_028830581.1 | XP_028686414.1 | 1488 | 85 | |||
XM_028830582.1 | XP_028686415.1 | 1366 | 85 | |||
XM_028830583.1 | XP_028686416.1 | 2926 | 85 | |||
GenBank mRNA | JU335429.1 | - | 7 - 8 | 1361 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
