Assay Details
Gene Symbol: PROX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260873.1 | NP_001247802.1 | 4 - 5 | 2152 | 74 | |
XM_015116188.2 | XP_014971674.1 | 2641 | 74 | |||
GenBank mRNA | JU320932.1 | - | 4 - 5 | 2152 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
