Assay Details
Gene Symbol: SLA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001086187.4 | XP_001086187.3 | 1671 | 87 | ||
XM_015146023.2 | XP_015001509.2 | 1398 | 87 | |||
XM_015146024.2 | XP_015001510.1 | 2152 | 87 | |||
XM_015146025.2 | XP_015001511.1 | 2052 | 87 | |||
XM_015146026.2 | XP_015001512.1 | 1947 | 87 | |||
XM_015146028.2 | XP_015001514.1 | 2303 | 87 | |||
XM_015146029.2 | XP_015001515.1 | 2048 | 87 | |||
XM_028852928.1 | XP_028708761.1 | 2019 | 87 | |||
XM_028852929.1 | XP_028708762.1 | 2047 | 87 | |||
XM_028852930.1 | XP_028708763.1 | 1776 | 87 | |||
GenBank mRNA | JU471466.1 | - | 680 | 87 | ||
JU471467.1 | - | 680 | 87 | |||
JU471468.1 | - | 680 | 87 | |||
JU471469.1 | - | 680 | 87 | |||
JU478226.1 | - | 719 | 87 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() ![]() |
Panther Classification:
