Assay Details
Gene Symbol: WDR83OS | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001194406.1 | NP_001181335.1 | 3 - 4 | 503 | 75 | |
GenBank mRNA | JU320875.1 | - | 3 - 4 | 412 | 75 | |
JU470489.1 | - | 3 - 4 | 347 | 75 | ||
JV043684.1 | - | 3 - 4 | 287 | 75 | ||
JV566224.1 | - | 3 - 4 | 504 | 75 | ||
JV639485.1 | - | 3 - 4 | 456 | 75 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() ![]() |
