Assay Details
Gene Symbol: EXTL3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002805291.3 | XP_002805337.2 | 3036 | 58 | ||
XM_015145080.2 | XP_015000566.1 | 2916 | 58 | |||
GenBank mRNA | JU333369.1 | - | 4 - 5 | 2871 | 58 | |
JU474661.1 | - | 3 - 4 | 2860 | 58 | ||
JV044888.1 | - | 4 - 5 | 2875 | 58 | ||
JV536498.1 | - | 4 - 5 | 2878 | 58 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
