Assay Details
Gene Symbol: NUAK1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001098986.3 | XP_001098986.2 | 1986 | 65 | ||
XM_028829824.1 | XP_028685657.1 | 1421 | 65 | |||
XM_028829825.1 | XP_028685658.1 | 301 | 65 | |||
GenBank mRNA | JV564655.1 | - | 4 - 5 | 494 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
