Assay Details
Gene Symbol: FKBP14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266094.1 | NP_001253023.1 | 3 - 4 | 568 | 115 | |
GenBank mRNA | JU321662.1 | - | 3 - 4 | 624 | 115 | |
JV045531.1 | - | 3 - 4 | 568 | 115 | ||
JV704667.1 | - | 3 - 4 | 627 | 115 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
