Assay Details
Gene Symbol: DISC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001114355.1 | NP_001107827.1 | 6 - 7 | 1639 | 107 | |
GenBank mRNA | AY681964.1 | - | 6 - 7 | 1639 | 107 |

More Information
Set Membership: |
![]() ![]() ![]() |
