Assay Details
Gene Symbol: PLPP5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001092746.4 | XP_001092746.3 | 103 | 102 | ||
XM_015145199.2 | XP_015000685.1 | 104 | 102 | |||
XM_015145200.2 | XP_015000686.1 | 104 | 102 |

More Information
Set Membership: |
![]() ![]() |
