Assay Details
Gene Symbol: RAB3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266792.2 | NP_001253721.1 | 3 - 4 | 436 | 95 | |
GenBank mRNA | JU321715.1 | - | 3 - 4 | 379 | 95 | |
JV489494.1 | - | 3 - 4 | 377 | 95 | ||
JV667301.1 | - | 3 - 4 | 436 | 95 |

More Information
Set Membership: |
![]() ![]() ![]() |
