Assay Details
Gene Symbol: OGA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015148051.2 | XP_015003537.1 | 933 | 120 | ||
XM_015148052.2 | XP_015003538.1 | 933 | 120 | |||
XM_028826799.1 | XP_028682632.1 | 933 | 120 | |||
GenBank mRNA | JU320291.1 | - | 2 - 3 | 661 | 120 | |
JU320292.1 | - | 2 - 3 | 661 | 120 | ||
JU472439.1 | - | 2 - 3 | 605 | 120 | ||
JV043778.1 | - | 2 - 3 | 666 | 120 | ||
JV705461.1 | - | 2 - 3 | 392 | 120 |

More Information
Set Membership: |
![]() ![]() ![]() |
