Assay Details
Gene Symbol: CDIPT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260643.1 | NP_001247572.1 | 1 - 2 | 441 | 95 | |
XM_028840493.1 | XP_028696326.1 | 921 | 95 | |||
GenBank mRNA | JU321684.1 | - | 1 - 2 | 445 | 95 | |
JU472849.1 | - | 1 - 2 | 434 | 95 | ||
JV044080.1 | - | 1 - 2 | 441 | 95 | ||
JV567513.1 | - | 1 - 2 | 449 | 95 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001260643.1, XM_028840493.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
