Assay Details
Gene Symbol: ENY2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001265903.1 | NP_001252832.1 | 1 - 2 | 95 | 88 | |
GenBank mRNA | JU473761.1 | - | 1 - 2 | 95 | 88 | |
JV528330.1 | - | 1 - 2 | 133 | 88 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001265903.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
