Assay Details
Gene Symbol: NT5C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001093693.4 | XP_001093693.4 | 339 | 93 | ||
GenBank mRNA | JU485347.1 | - | 1 - 2 | 169 | 93 | |
JV046901.1 | - | 1 - 2 | 195 | 93 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() |
