Assay Details
Gene Symbol: MAX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001261369.2 | NP_001248298.1 | 2 - 3 | 316 | 77 | |
XM_015143840.2 | XP_014999326.1 | 523 | 77 | |||
XM_028850344.1 | XP_028706177.1 | 496 | 77 | |||
GenBank mRNA | JU320631.1 | - | 3 - 4 | 339 | 77 | |
JU320632.1 | - | 2 - 3 | 312 | 77 | ||
JU472565.1 | - | 2 - 3 | 319 | 77 | ||
JU472566.1 | - | 3 - 4 | 346 | 77 | ||
JV043408.1 | - | 3 - 4 | 344 | 77 | ||
JV043409.1 | - | 2 - 3 | 317 | 77 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
This assay is in the 5'UTR region of the XM_028850344.1 transcripts.
This assay is in the 5'UTR region of the XM_028850344.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
