Assay Details
Gene Symbol: TOGARAM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015143616.2 | XP_014999102.2 | 5317 | 143 | ||
XM_028851520.1 | XP_028707353.1 | 5320 | 143 | |||
XM_028851522.1 | XP_028707355.1 | 5221 | 143 | |||
XM_028851523.1 | XP_028707356.1 | 5017 | 143 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() |
