Assay Details
Gene Symbol: CDK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001113345.4 | XP_001113345.2 | 588 | 58 | ||
XM_015151938.2 | XP_015007424.1 | 591 | 58 | |||
GenBank mRNA | JU320958.1 | - | 3 - 4 | 530 | 58 | |
JU473899.1 | - | 3 - 4 | 554 | 58 | ||
JV045857.1 | - | 3 - 4 | 556 | 58 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
