Assay Details
Gene Symbol: BAG3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266057.2 | NP_001252986.1 | 1 - 2 | 478 | 51 | |
XM_015148170.2 | XP_015003656.2 | 495 | 51 | |||
GenBank mRNA | JU337213.1 | - | 1 - 2 | 195 | 51 | |
JU477927.1 | - | 1 - 2 | 491 | 51 | ||
JV048770.1 | - | 1 - 2 | 482 | 51 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() ![]() |
