Assay Details
Gene Symbol: WNT4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001100814.4 | XP_001100814.1 | 500 | 66 | ||
XM_028838318.1 | XP_028694151.1 | 500 | 66 | |||
GenBank mRNA | JV654085.1 | - | 1 - 2 | 231 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
