Assay Details
Gene Symbol: LOC714689 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015148074.2 | XP_015003560.2 | 679 | 123 | ||
XM_015148075.2 | XP_015003561.2 | 679 | 123 | |||
XM_015148076.2 | XP_015003562.2 | 622 | 123 | |||
XM_015148077.2 | XP_015003563.2 | 666 | 123 | |||
XM_015148078.2 | XP_015003564.2 | 597 | 123 | |||
GenBank mRNA | JU470889.1 | - | 6 - 7 | 524 | 123 |
Gene Symbol: NFKB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028825898.1 | XP_028681731.1 | 640 | 123 | ||
GenBank mRNA | JV062003.1 | - | 6 - 7 | 539 | 123 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
