Assay Details
Gene Symbol: DMWD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_028839492.1 | XP_028695325.1 | 1984 | 88 | ||
GenBank mRNA | JV046347.1 | - | 3 - 4 | 1913 | 88 | |
JV667355.1 | - | 3 - 4 | 1909 | 88 | ||
JV708584.1 | - | 3 - 4 | 1731 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() |
