Assay Details
Gene Symbol: CAMP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001033509.2 | NP_001028681.1 | 1 - 2 | 209 | 110 | |
GenBank mRNA | AF181954.1 | - | 1 - 2 | 212 | 110 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
