Assay Details
Gene Symbol: IL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001032904.1 | NP_001028076.1 | 183 | 71 | ||
GenBank mRNA | AF457197.1 | - | 47 | 71 | ||
AY376144.1 | - | 183 | 71 | |||
EF173856.1 | - | 53 | 71 | |||
L26027.1 | - | 183 | 71 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
