Assay Details
Gene Symbol: TRIM5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001032910.1 | NP_001028082.1 | 2 - 3 | 698 | 119 | |
GenBank mRNA | AY523632.1 | - | 2 - 3 | 698 | 119 | |
AY523633.1 | - | 1 - 2 | 428 | 119 | ||
AY625001.1 | - | 1 - 2 | 428 | 119 | ||
DQ842020.1 | - | 1 - 2 | 428 | 119 | ||
DQ842021.1 | - | 1 - 2 | 428 | 119 | ||
EF113914.1 | - | 1 - 2 | 428 | 119 | ||
EF113915.1 | - | 1 - 2 | 428 | 119 | ||
EF113916.1 | - | 1 - 2 | 428 | 119 | ||
EF113917.1 | - | 1 - 2 | 428 | 119 | ||
EF113918.1 | - | 1 - 2 | 428 | 119 | ||
EF113919.1 | - | 1 - 2 | 428 | 119 | ||
JU336825.1 | - | 2 - 3 | 549 | 119 | ||
JU477573.1 | - | 2 - 3 | 586 | 119 | ||
JU477574.1 | - | 2 - 3 | 586 | 119 | ||
JU477575.1 | - | 2 - 3 | 586 | 119 | ||
JU477576.1 | - | 2 - 3 | 586 | 119 | ||
JU477577.1 | - | 2 - 3 | 586 | 119 | ||
JV048518.1 | - | 2 - 3 | 559 | 119 | ||
JX865253.1 | - | 1 - 2 | 428 | 119 | ||
JX865254.1 | - | 1 - 2 | 428 | 119 |

More Information
Set Membership: |
![]() ![]() ![]() |
