Assay Details
Gene Symbol: APEX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001258109.1 | NP_001245038.1 | 2 - 3 | 266 | 96 | |
XM_015143085.2 | XP_014998571.1 | 512 | 96 | |||
XM_015143087.2 | XP_014998573.1 | 530 | 96 | |||
XM_015143088.2 | XP_014998574.1 | 516 | 96 | |||
XM_015143089.2 | XP_014998575.1 | 506 | 96 | |||
XM_015143090.2 | XP_014998576.1 | 400 | 96 | |||
XM_015143091.2 | XP_014998577.1 | 404 | 96 | |||
GenBank mRNA | JU326201.1 | - | 1 - 2 | 182 | 96 | |
JU475401.1 | - | 2 - 3 | 266 | 96 | ||
JV046924.1 | - | 3 - 4 | 502 | 96 | ||
JV046925.1 | - | 3 - 4 | 436 | 96 | ||
JV046926.1 | - | 2 - 3 | 311 | 96 |

More Information
Set Membership: |
![]() ![]() ![]() |
