Assay Details
Gene Symbol: MRPL23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266484.1 | NP_001253413.1 | 1 - 2 | 34 | 82 | |
GenBank mRNA | JV047869.1 | - | 1 - 2 | 34 | 82 | |
JV602493.1 | - | 1 - 2 | 115 | 82 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001266484.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
