Assay Details
Gene Symbol: FKBP14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001266094.1 | NP_001253023.1 | 1 - 2 | 285 | 98 | |
GenBank mRNA | JU321662.1 | - | 1 - 2 | 341 | 98 | |
JV045531.1 | - | 1 - 2 | 285 | 98 | ||
JV704667.1 | - | 1 - 2 | 344 | 98 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
Set Membership: |
![]() ![]() ![]() |
