Assay Details
Gene Symbol: TRIQK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001265814.1 | NP_001252743.1 | 2 - 3 | 286 | 65 | |
XM_015145636.2 | XP_015001122.1 | 296 | 65 | |||
GenBank mRNA | JV045323.1 | - | 2 - 3 | 286 | 65 |

More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001265814.1, XM_015145636.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
