Assay Details
Gene Symbol: KBTBD4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001260536.1 | NP_001247465.1 | 1 - 2 | 92 | 60 | |
GenBank mRNA | JV047607.1 | - | 1 - 2 | 92 | 60 |

More Information
Additional Information:
This assay maps with 100% identity to the human homologue, and the human gene symbol was added in the Alias field to assist with searches.
This assay is in the 5'UTR region of the NM_001260536.1 transcripts.
This assay is in the 5'UTR region of the NM_001260536.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
