Assay Details
Gene Symbol: TRIM13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002800768.3 | XP_002800814.2 | 505 | 72 | ||
XM_015121091.2 | XP_014976577.2 | 212 | 72 | |||
XM_015121092.2 | XP_014976578.2 | 288 | 72 | |||
XM_015121093.2 | XP_014976579.2 | 146 | 72 | |||
XM_028837505.1 | XP_028693338.1 | 232 | 72 | |||
GenBank mRNA | JU478625.1 | - | 2 - 3 | 211 | 72 | |
JU478627.1 | - | 1 - 2 | 744 | 72 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_002800768.3, XM_015121091.2, XM_015121092.2, XM_015121093.2, XM_028837505.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
